Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Methyl-2-piperidnyl)methanamine, 97%, Thermo Scientific™

Catalog No. MO01154DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO01154DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01154DA Supplier Thermo Scientific Chemicals Supplier No. MO01154DA
Add to Cart

Chemical Identifiers

CAS 5298-72-6
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD05022432
InChI Key PPUMJZMVFCLQBI-UHFFFAOYNA-N
PubChem CID 2794677
SMILES CN1CCCCC1CN

Specifications

Density 0.9102g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 1-methylpiperidin-2-yl methanamine, 1-methyl-2-piperidinyl methanamine, 2-piperidinemethanamine, 1-methyl, 1-1-methylpiperidin-2-yl methanamine, c-1-methyl-piperidin-2-yl-methylamine, 1-methyl-2-piperidnyl methanamine, 1-methyl-2-piperidyl methylamine, pubchem8033, 1-methyl-2-aminomethylpiperidine, 2-piperidinemethanamine,1-methyl
IUPAC Name 1-(1-methylpiperidin-2-yl)methanamine
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material (1-Methyl-2-piperidinyl)methanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.