Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Methyl-2-Propenylmagnesium Chloride Solution

Catalog No. 420042100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-004-2100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 42-004-2100ML Supplier Sigma Aldrich Supplier No. 420042100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Methyl-2-propenylmagnesium chloride is a Grignard type allylating reagent that can be used for the preparation of corresponding homoallylic alcohols by reacting with various aldehydes. It can also be utilized in the synthesis of 3,4-dihydro-4-(1-methyl-2-propen-1-yl)-2(1H)-pyrimidinethione derivatives by treating with 2-mercaptopyrimidine.

Specifications

CAS 119904-90-4
Molecular Formula C7H14N2 -+ 2HCl
Linear Formula C7H14N2 -+ 2HCl
MDL Number MFCD00191753
Quantity 100 mL
Molecular Weight (g/mol) 199.12 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.