Learn More
Sigma Aldrich Fine Chemicals Biosciences 1 Methyl 2 pyrrolidinone b
Catalog No. 502902873
Encompass
N-Methyl-2-pyrrolidinone (N-methylpyrrolidone NMP) is a polar cyclic organic solvent. A study on the impact of NMP on Saccharomyces cerevisiae revealed that it induced aneuploidy. Characteristic properties of NMP include low viscosity excellent dissolving power and enhanced demulsifying property. It is a better alternative to phenol in selectively treating lube feedstock. The behavior of fullerene-C60 in NMP SOLUTIONon has been examined based on electronic absorption and photoluminescence spectral data. A SOLUTIONon of sodium borohydride (NaBH4) in NMP showed increased reactivity towards debromination of alkyl bromide. The enhanced selectivity of NMP-ethylene glycol solvent mixture for aromatic hydrocarbon extraction has been analyzed by gas chromatography.
Catalog No. 50-290-2873 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4944962L
N-Methyl-2-pyrrolidinone (N-methylpyrrolidone NMP) is a polar cyclic organic solvent. A study on the impact of NMP on Saccharomyces cerevisiae revealed that it induced aneuploidy. Characteristic properties of NMP include low viscosity excellent dissolving power and enhanced demulsifying property. It is a better alternative to phenol in selectively treating lube feedstock. The behavior of fullerene-C60 in NMP SOLUTIONon has been examined based on electronic absorption and photoluminescence spectral data. A SOLUTIONon of sodium borohydride (NaBH4) in NMP showed increased reactivity towards debromination of alkyl bromide. The enhanced selectivity of NMP-ethylene glycol solvent mixture for aromatic hydrocarbon extraction has been analyzed by gas chromatography.