Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-2-pyrrolidinone, CHROMASOLV™ Plus, for HPLC, ≥99%, Solstice

Catalog No. 6002238 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 4 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-022-38 2.5 L Glass Bottle
60-022-37 1 L Glass Bottle
60-045-740 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 60-022-38 Supplier Solstice Supplier No. 2704582.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™ Plus, for HPLC,≥99%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Melting Point -24°C (literature)
Absorbance max. 0.01 at 400 nm, max. 0.01 at 350 nm, max. 0.10 at 325 nm, max. 0.50 at 300 nm, max. 1.0 at 285 nm
CAS Max % 100.0000%
Density 1.028 g/mL at 25 °C(lit.)
Quantity 2.5 L
Flash Point 91°C
Packaging Glass Bottle
Linear Formula C5H9NO
Beilstein 106420
Boiling Point 204°C
Formula Weight 99.13
Percent Purity >99%
Grade HPLC
Chemical Name or Material 1-Methyl-2-pyrrolidinone, CHROMASOLV™ Plus, for HPLC, ≥99%
Show More Show Less
ShelfLife 1980 days from date of manufacture
EINECSNumber 212-828-1
Recommended Storage Room Temp
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.