Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-2-pyrrolidinone, Solstice™

Catalog No. 60090039 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Returnable Stainless Steel Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-039 20 L Returnable Stainless Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-039 Supplier Solstice Supplier No. 44377820L

missing translation for 'validMainframeMsg'

ACS Reagent, ≥99.0%

  • Low UV impurity profile
  • TRUSTED_SUSTAINABILITY

    Chemical Identifiers

    CAS 872-50-4
    PubChem CID 13387
    ChEBI CHEBI:7307

    Specifications

    Quantity 20 L
    Packaging Returnable Stainless Steel Can
    Physical Form Liquid
    Percent Purity min. 99.0 %
    Grade ACS
    Recommended Storage Room Temperature
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.