Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-2-Pyrrolidone, 99.7%, MilliporeSigma™

Catalog No. 50947960 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-960 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-947-960 Supplier MilliporeSigma Supplier No. MX13921
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Melting Point −24°C
pH 8.5 to 10.0 (100 g/L, H2O, 20°C)
Boiling Point 202°C (1013 hPa)
Flash Point 91°C
Assay Percent Range 99.7% min
Packaging Glass Bottle, Pre-Packed
Quantity 4 L
Vapor Pressure 0.32 hPa (20°C)
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.