Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-2-pyrrolidone, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18608120 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
20 L
200 L
Packaging:
Drum
Steel Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-120 20 L Steel Can
18-608-121 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-608-120 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M128020LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 872-50-4
Molecular Weight (g/mol) 99.14

Specifications

CAS Max % 1
Quantity 20 L
Packaging Steel Can
Water 0.0005
Boiling Point 202°C
Percent Purity 0.99
Grade Reagent
Chemical Name or Material 1-Methyl-2-pyrrolidone
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.