Learn More
Description
- Every lot is tested to ensure interference free baselines for interference free determination of Class 1, 2, and 3 residual solvents
- 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
- 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
- Processed for low water content to facilitate the extraction of organic volatile impurities, or VOC
Specifications
Specifications
| Absorbance | 0.100 AU Maximum (325nm), 0.030 AU Maximum (350nm), 0.010 AU Maximum (400nm) |
| Color | Colorless to Yellow |
| Quantity | 1 L |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Water | 0.05% Maximum |
| Formula Weight | 99.13 |
| Percent Purity | ≥99.9% |
| Grade | GC Headspace |
| Show More |
Safety and Handling
| Recommended Storage | Room temperature, packaged under inert atmosphere. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.