Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3,5-dinitro-2-pyridone 98.0+%, TCI America™
SDP

Catalog No. M28201G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M28201G Supplier TCI America Supplier No. M28201G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14150-94-8
Molecular Formula C6H5N3O5
Molecular Weight (g/mol) 199.12
MDL Number MFCD00456280
InChI Key QARVELJVEBLWGK-UHFFFAOYSA-N
Synonym 1-Methyl-3,5-dinitropyridin-2(1H)-one
PubChem CID 3621236
IUPAC Name 1-methyl-3,5-dinitro-1,2-dihydropyridin-2-one
SMILES CN1C=C(C=C(C1=O)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 176°C
Color Yellow
Quantity 1 g
Formula Weight 199.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-3,5-dinitro-2-pyridone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.