Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Methyl-3-Octylimidazolium Chloride

Catalog No. 958035G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
95-803-5G 5 g
95-803-50G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 95-803-5G Supplier Sigma Aldrich Supplier No. 958035G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Methyl-3-octylimidazolium chloride ionic liquid ([MOIM]Cl) can be used as: A reaction medium for the hydrolysis of sucrose to form hydroxymethylfurfural in the presence of HCl and metal chloride catalyst A structure-directing agent during the preparation of organized mesoporous alumina.Lipophilic ionic liquid

Specifications

CAS 64697-40-1
Density 1.01 g/mL at 20°C
Molecular Formula C12H23ClN2
Refractive Index n20/D 1.505-1.515
Linear Formula C12H23ClN2
Quantity 5 g
MDL Number MFCD03095432
Synonym OMIMCl
Molecular Weight (g/mol) 230.78 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.