Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-piperazinone, 95%, Thermo Scientific Chemicals

Catalog No. AAH5763303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5763303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5763303 Supplier Thermo Scientific Chemicals Supplier No. H5763303
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 34770-60-0 | Molecular Formula: C5H10N2O | Molecular Weight: 114.15 g/mol

It is a useful piperazine and also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful piperazine and also used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34770-60-0
Molecular Formula C5H10N2O
Molecular Weight (g/mol) 114.15
MDL Number MFCD11870124
InChI Key KVIZTDNKHOCNAM-UHFFFAOYSA-N
Synonym 1-methyl-3-oxopiperazine, 1-methyl-3-piperazinone, 1-methylpiperazin-3-one, piperazinone, 4-methyl, pubchem23318, 1-methyl-3-piperizinone, 4-methyl-2-piperazinone, 4-methyl-piperazin-2-one, 2-piperazinone,4-methyl
PubChem CID 13704283
IUPAC Name 4-methylpiperazin-2-one
SMILES CN1CCNC(=O)C1

Specifications

Melting Point 84°C to 90°C
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 114.15
Percent Purity 95%
Chemical Name or Material 1-Methyl-3-piperazinone
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.