Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-propylimidazolium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. M30365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30365G 5 g
M303625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30365G Supplier TCI America Supplier No. M30365G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 244193-48-4
Molecular Formula C7H13BF4N2
Molecular Weight (g/mol) 211.999
MDL Number MFCD14156016
InChI Key XFTQQJUVYZPNRC-UHFFFAOYSA-N
PubChem CID 11171853
IUPAC Name 1-methyl-3-propylimidazol-1-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CCCN1C=C[N+](=C1)C

Specifications

Melting Point 17°C
Color Yellow
Quantity 5 g
UN Number 1760
Formula Weight 212.00
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-propylimidazolium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.