Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride, 97+%, Thermo Scientific™

Catalog No. AW00087CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AW00087CB 250 mg Amber Glass Bottle
AW00087DA 1 g Amber Glass Bottle
AW00087DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AW00087CB Supplier Thermo Scientific Chemicals Supplier No. AW00087CB
Add to Cart

Chemical Identifiers

CAS 519056-67-8
Molecular Formula C5H4ClF3N2O2S
Molecular Weight (g/mol) 248.604
InChI Key HJFWSNIHZZKYIK-UHFFFAOYSA-N
Synonym 1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl pyrazole-4-sulfonyl chloride, 1h-pyrazole-4-sulfonylchloride,1-methyl-3-trifluoromethyl, chloro 1-methyl-3-trifluoromethyl pyrazol-4-yl sulfone, 1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonylchloride, 1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl
PubChem CID 2794561
IUPAC Name 1-methyl-3-(trifluoromethyl)pyrazole-4-sulfonyl chloride
SMILES CN1C=C(C(=N1)C(F)(F)F)S(=O)(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 248.61
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.