Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-4-(4-piperidinyl)piperazine, 98%

Catalog No. AAH2740806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2740806 5 g
AAH2740803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2740806 Supplier Thermo Scientific Chemicals Supplier No. H2740806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 53617-36-0 | C10H21N3 | 183.30 g/mol

It is used both as a reagent and building block in several synthetic applications. As intermediate. As an excellent catalyst for many condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used both as a reagent and building block in several synthetic applications. As intermediate. As an excellent catalyst for many condensation reactions.

Solubility
Soluble in water.

Notes
Air Sensitive & Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water, moisture, acids. Store under dry inert gas. Refrigerated (2-4°C).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53617-36-0
Molecular Formula C10H21N3
Molecular Weight (g/mol) 183.30
MDL Number MFCD03274729
InChI Key MRYYJGQKVGZGSB-UHFFFAOYSA-N
Synonym 1-methyl-4-piperidin-4-yl piperazine, 1-methyl-4-4-piperidinyl piperazine, 1-methyl-4-piperidin-4-yl-piperazine, piperazine, 1-methyl-4-4-piperidinyl, 4-methyl-1-4-piperidyl piperazine, acmc-1b13b, 1-methyl-4-4-piperidyl piperazine, 1-methyl-4-piperidin-4yl-piperazine, 1-methyl-4-4-piperidino piperazine
PubChem CID 795707
SMILES CN1CCN(CC1)C1CCNCC1

Specifications

Melting Point 53°C to 56°C
Quantity 5 g
UN Number UN3259
Sensitivity Air sensitive; Hygroscopic
Solubility Information Soluble in water.
IUPAC Name 1-methyl-4-piperidin-4-ylpiperazine
Formula Weight 183.3
Percent Purity 98%
Chemical Name or Material 1-Methyl-4-(4-piperidinyl)piperazine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.