Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-methyl-4-imidazoleacetic acid hydrochloride | 35454-39-8 | MFCD00036998 | 99.3% | 176.60 | C6H9ClN2O2 | 25 MG
SDP

Catalog No. 5000209045
Encompass_Preferred

1-methyl-4-imidazoleacetic acid hydrochloride is the hydrochloride salt of a stable histamine metabolite used as a biochemical research reagent and analytical reference for studies of histamine metabolism. It is supplied as a solid in small pack sizes suitable for laboratory use.

  • Chemical name: 1-methyl-4-imidazoleacetic acid hydrochloride.
  • Cas number: 35454-39-8.
  • Molecular formula: C6H9ClN2O2.
  • Molecular weight: 176.60 g/mol.
  • Purity: 99.25% (analytical grade).
  • Physical form: solid.
  • Available pack sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.
  • Also available as 10 mM 1 mL solution in DMSO for immediate use.

Catalog No. 50-002-09045 Supplier Medchemexpress LLC Supplier No. 25MGHYW060470
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-methyl-4-imidazoleacetic acid hydrochloride is the hydrochloride salt of a stable histamine metabolite used as a biochemical research reagent and analytical reference for studies of histamine metabolism. It is supplied as a solid in small pack sizes suitable for laboratory use.

  • Chemical name: 1-methyl-4-imidazoleacetic acid hydrochloride.
  • Cas number: 35454-39-8.
  • Molecular formula: C6H9ClN2O2.
  • Molecular weight: 176.60 g/mol.
  • Purity: 99.25% (analytical grade).
  • Physical form: solid.
  • Available pack sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.
  • Also available as 10 mM 1 mL solution in DMSO for immediate use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.