Learn More
Medchemexpress LLC 1-methyl-5-(2'-methyl-[1,1'-biphenyl]-4-yl)-1H-benzo[d]triazole-7-carboxylic acid | 1623416-31-8 | MFCD33023454 | 98.3% | C21H17N3O2 | 10MG
SDP
Catalog No. 5000258405
Encompass_Preferred
AG-636 is a potent, reversible, selective, and orally active dihydroorotate dehydrogenase (DHODH) inhibitor with an IC50 of 17 nM. It is supplied as a high-purity research reagent for biochemical and cellular studies investigating pyrimidine biosynthesis and anticancer pathways.
  • Potent DHODH inhibition (IC50 17 nM).
  • Reversible and orally active mechanism of action.
  • High purity for research applications.
  • Molecular formula C21H17N3O2.
  • Available as a small research pack for laboratory use.
Catalog No. 50-002-58405 Supplier Medchemexpress LLC Supplier No. HY13746310MG
AG-636 is a potent, reversible, selective, and orally active dihydroorotate dehydrogenase (DHODH) inhibitor with an IC50 of 17 nM. It is supplied as a high-purity research reagent for biochemical and cellular studies investigating pyrimidine biosynthesis and anticancer pathways.
  • Potent DHODH inhibition (IC50 17 nM).
  • Reversible and orally active mechanism of action.
  • High purity for research applications.
  • Molecular formula C21H17N3O2.
  • Available as a small research pack for laboratory use.