Learn More
Medchemexpress LLC 1-Methyl-5-mercapto-1,2,3,4-tetrazole-d3 | 345909-96-8 | 99.9% | 119.16 | 10 MG
SDP
Catalog No. 5000451815
Encompass_Preferred
1-Methyl-5-mercapto-1,2,3,4-tetrazole-d3 is a deuterium-labeled stable isotope of 1-Methyl-1H-tetrazole-5-thiol. It is intended for research use only and can be utilized as a tracer or an internal standard for quantitative analysis. Deuterium substitution in drug molecules is being studied for its potential impact on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-51815 Supplier Medchemexpress LLC Supplier No. HYW010517S10MG
Add to Cart
Add to Cart
1-Methyl-5-mercapto-1,2,3,4-tetrazole-d3 is a deuterium-labeled stable isotope of 1-Methyl-1H-tetrazole-5-thiol. It is intended for research use only and can be utilized as a tracer or an internal standard for quantitative analysis. Deuterium substitution in drug molecules is being studied for its potential impact on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.