Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-methyl-5-phenyl-1H-pyrazol-4-yl)methanol, Thermo Scientific™

Catalog No. CC28109DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC28109DE 5 g
CC28109CB 250 mg
CC28109DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC28109DE Supplier Thermo Scientific Chemicals Supplier No. CC28109DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 499785-47-6
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD04115390
InChI Key ZVGZXURABHDVPS-UHFFFAOYSA-N
Synonym 1-methyl-5-phenyl-1h-pyrazol-4-yl methanol, 1-methyl-5-phenylpyrazol-4-yl methanol, 1-methyl-5-phenylpyrazol-4-yl methan-1-ol, bck
PubChem CID 2795476
SMILES CN1N=CC(CO)=C1C1=CC=CC=C1

Specifications

Packaging Amber glass bottle
Quantity 5 g
IUPAC Name (1-methyl-5-phenylpyrazol-4-yl)methanol
Formula Weight 188.23
Percent Purity 97%
Chemical Name or Material (1-methyl-5-phenyl-1H-pyrazol-4-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.