Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-methyl-5-phenyl-1h-pyrazole-3-carbaldehyde, 97%, Thermo Scientific™

Catalog No. CC46504CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC46504CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC46504CB Supplier Thermo Scientific Chemicals Supplier No. CC46504CB

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 124344-94-1
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD08271935
InChI Key AGEKFZHCMVDIFA-UHFFFAOYSA-N
Synonym 1-methyl-5-phenyl-1h-pyrazole-3-carbaldehyde, 3-formyl-1-methyl-5-phenyl-1h-pyrazole, 1-methyl-5-phenylpyrazole-3-carboxaldehyde, 1h-pyrazole-3-carboxaldehyde,1-methyl-5-phenyl, acmc-20e3zm, 3-formyl-1-methyl-1h-pyrazol-5-yl benzene
PubChem CID 14354446
IUPAC Name 1-methyl-5-phenylpyrazole-3-carbaldehyde
SMILES CN1C(=CC(=N1)C=O)C2=CC=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 1-Methyl-5-phenyl-1H-pyrazole-3-carbaldehyde
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.