Learn More
eMolecules​ 170682-50-5 | 1-Methyl-5-(tributylstannyl)-1H-pyrazole | Synthonix - Stock | MFCD19686983 | 371.156 | C16H32N2Sn | 95.000 | CCCC[Sn](CCCC)(CCCC)c1ccnn1C | 5g | 525921665
Catalog No. 502696640 Shop All eMolecules Inc. Products
Encompass_Preferred
1-Methyl-5-(tributylstannyl)-1H-pyrazole | Synthonix - Stock | 170682-50-5 | MFCD19686983 | 371.156 | C16H32N2Sn | 95.000 | CCCC[Sn](CCCC)(CCCC)c1ccnn1C | 5g | 525921665
Catalog No. 50-269-6640 Supplier eMolecules​ Supplier No. 525921665
1-Methyl-5-(tributylstannyl)-1H-pyrazole | Synthonix - Stock | 170682-50-5 | MFCD19686983 | 371.156 | C16H32N2Sn | 95.000 | CCCC[Sn](CCCC)(CCCC)c1ccnn1C | 5g | 525921665