Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. M2753200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2753200MG 200 mg
M27531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2753200MG Supplier TCI America Supplier No. M2753200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 119083-00-0
Molecular Formula C6H5F3N2O2
Molecular Weight (g/mol) 194.113
MDL Number MFCD04110182
InChI Key VAKOSNKAXYJZRG-UHFFFAOYSA-N
PubChem CID 2807771
IUPAC Name 1-methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid
SMILES CN1C(=C(C=N1)C(=O)O)C(F)(F)F

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 200 mg
Formula Weight 194.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.