Learn More
Medchemexpress LLC 1-(Methyl-d3)piperazine | 1093380-08-5 | 99.2% | C5H9D3N2 | 25 MG
SDP
Catalog No. 5000357641
Encompass_Preferred
1-(Methyl-d3)piperazine is the deuterium labeled N-Methylpiperazine. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, which involves incorporating stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Influences pharmacokinetic and metabolic profiles of drugs through deuteration
Catalog No. 50-003-57641 Supplier Medchemexpress LLC Supplier No. 25MGHY78871S2
1-(Methyl-d3)piperazine is the deuterium labeled N-Methylpiperazine. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, which involves incorporating stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Influences pharmacokinetic and metabolic profiles of drugs through deuteration