Learn More
Medchemexpress LLC HY-W017006 10mg Medchemexpress, 1-Methyl-L-histidine CAS:332-80-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW01700610MG
Encompass_Preferred
Medchemexpress, HY-W017006 10mg 1-Methyl-L-histidine CAS:332-80-9 1-Methyl-L-histidine is an objective indicator of meat ingestion and exogenous 3-methylhistidine (3MH) intake. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4624