Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl N-(tert-Butoxycarbonyl)-L-aspartate 97.0+%, TCI America™
SDP

Catalog No. M29705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M29705G Supplier TCI America Supplier No. M29705G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 98045-03-5
Molecular Formula C10H17NO6
Molecular Weight (g/mol) 247.247
MDL Number MFCD03094777
InChI Key IWFIVTBTZUCTQH-LURJTMIESA-N
Synonym boc-asp-ome, 1-methyl n-boc-l-aspartate, n-boc-l-aspartic acid 1-methyl ester, 3s-3-tert-butoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, n-boc asp oh ome, 1-methyl n-tert-butoxycarbonyl-l-aspartate, n-tert-butoxycarbonylaspartic acid a-methyl ester, n-tert-butoxycarbonyl aspartic acid 1-methyl ester, n-tert-butoxycarbonyl-l-aspartic acid 1-methyl est, n-tert-butoxycarbonyl-l-aspartic acid 1-methyl ester
PubChem CID 11021202
IUPAC Name (3S)-4-methoxy-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OC

Specifications

Melting Point 89°C
Color White
Quantity 5 g
Formula Weight 247.25
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl N-(tert-Butoxycarbonyl)-L-aspartate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.