Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-methyl-nicotinamide iodide | 6456-44-6 | MFCD00031898 | 264.06 | 250 MG
SDP

Catalog No. 5000366414
Encompass_Preferred

TRIA-662 iodide is the iodide form of TRIA-662, an endogenous metabolite that possesses antithrombotic and anti-inflammatory activities. This product is for research use only and not for human use. It is a nicotinic acid derivative known for its dopaminergic neurotoxicity.

  • Also known as 1-methylnicotinamide iodide
  • Classified as an endogenous metabolite
  • Molecular formula: C7H9IN2O
  • Appears as an off-white to light yellow solid powder
  • Recommended storage: powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month
  • Stable at room temperature for shipping
  • High solubility in DMSO (approx. 66.67 mg/mL)
  • In vivo solubility of at least 5 mg/mL in 10% DMSO + 90% (20% SBE-β-CD in Saline)

Catalog No. 50-003-66414 Supplier Medchemexpress LLC Supplier No. 250MGHYW009418
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TRIA-662 iodide is the iodide form of TRIA-662, an endogenous metabolite that possesses antithrombotic and anti-inflammatory activities. This product is for research use only and not for human use. It is a nicotinic acid derivative known for its dopaminergic neurotoxicity.

  • Also known as 1-methylnicotinamide iodide
  • Classified as an endogenous metabolite
  • Molecular formula: C7H9IN2O
  • Appears as an off-white to light yellow solid powder
  • Recommended storage: powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month
  • Stable at room temperature for shipping
  • High solubility in DMSO (approx. 66.67 mg/mL)
  • In vivo solubility of at least 5 mg/mL in 10% DMSO + 90% (20% SBE-β-CD in Saline)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.