Learn More
Medchemexpress LLC HY-113306 5mg Medchemexpress, 1-Methyladenine CAS:5142-22-3 Purity:>98%
SDP
Catalog No. 501969240
Encompass_Preferred
Medchemexpress, HY-113306 5mg 1-Methyladenine CAS:5142-22-3 1-Methyladenine is a product of alkylation damage in DNA which can be repaired by damage reversal by oxidative demethylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9240 Supplier Medchemexpress LLC Supplier No. HY1133065MG
Medchemexpress, HY-113306 5mg 1-Methyladenine CAS:5142-22-3 1-Methyladenine is a product of alkylation damage in DNA which can be repaired by damage reversal by oxidative demethylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.