Learn More
Medchemexpress LLC 1-methyladenosine | 15763-06-1 | MFCD00038421 | 99.9% | 281.27 g/mol | C11H15N5O4 | 5 MG
SDP
Catalog No. 5000262474
Encompass_Preferred
1-methyladenosine is an endogenous RNA nucleoside modification provided as an analytical standard for research and analytical applications, particularly studies of RNA methylation and cancer-related biomarker analysis. Molecular formula C11H15N5O4; molecular weight 281.27 g/mol.
  • Analytical standard for RNA methylation and biomarker studies.
  • High purity (>99.9% by HPLC) suitable for analytical workflows.
  • Available in solid and solution formats for flexible use.
  • Identity confirmed by spectrometric analysis and certificate of analysis.
  • Suitable for research in cancer biology and metabolic pathway investigation.
Catalog No. 50-002-62474 Supplier Medchemexpress LLC Supplier No. HY113081R5MG
1-methyladenosine is an endogenous RNA nucleoside modification provided as an analytical standard for research and analytical applications, particularly studies of RNA methylation and cancer-related biomarker analysis. Molecular formula C11H15N5O4; molecular weight 281.27 g/mol.
  • Analytical standard for RNA methylation and biomarker studies.
  • High purity (>99.9% by HPLC) suitable for analytical workflows.
  • Available in solid and solution formats for flexible use.
  • Identity confirmed by spectrometric analysis and certificate of analysis.
  • Suitable for research in cancer biology and metabolic pathway investigation.