Learn More
Medchemexpress LLC 1-methyladenosine-d3 | 00-00-0 | 284.29 | C11H12D3N5O4 | 5 MG
SDP
Catalog No. 5000262422
Encompass_Preferred
1-Methyladenosine-d3 is the deuterium-labeled form of 1-methyladenosine supplied as a stable isotope research standard. It is used as an internal standard and tracer in LC-MS/MS and metabolic studies of RNA modifications and nucleoside metabolism. The product is provided with supporting documentation for analytical use.
  • Deuterium-labeled internal standard for quantitative mass spectrometry.
  • Suitable for LC-MS/MS method development and validation.
  • Supports studies of RNA modification and nucleoside metabolism.
  • Supplied with datasheet, SDS, and certificate of analysis.
  • Available in small-mass quantities for analytical use.
Catalog No. 50-002-62422 Supplier Medchemexpress LLC Supplier No. HY113081S5MG
1-Methyladenosine-d3 is the deuterium-labeled form of 1-methyladenosine supplied as a stable isotope research standard. It is used as an internal standard and tracer in LC-MS/MS and metabolic studies of RNA modifications and nucleoside metabolism. The product is provided with supporting documentation for analytical use.
  • Deuterium-labeled internal standard for quantitative mass spectrometry.
  • Suitable for LC-MS/MS method development and validation.
  • Supports studies of RNA modification and nucleoside metabolism.
  • Supplied with datasheet, SDS, and certificate of analysis.
  • Available in small-mass quantities for analytical use.