Learn More
Medchemexpress LLC 1-Methyladenosine-d3 hydriodide | 15763-06-1 | 98.8% | 412.20 | 1 MG
SDP
Catalog No. 5000436472
Encompass_Preferred
1-Methyladenosine-d3 hydriodide is a deuterium-labeled version of 1-Methyladenosine. This RNA modification serves as a tumor marker, with elevated levels linked to cancer development. Its methylation leads to the upregulation of PPARδ expression, regulating cholesterol metabolism and activating the Hedgehog signaling pathway, contributing to liver tumorigenesis. This compound is intended for research use only and can be used as a tracer and internal standard for quantitative analysis.
  • Deuterium-labeled version of 1-Methyladenosine
  • Functions as a tumor marker
  • Upregulates PPARδ expression
  • Regulates cholesterol metabolism
  • Activates Hedgehog signaling pathway
  • Contributes to liver tumorigenesis
  • Suitable as a tracer for quantitative analysis
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-36472 Supplier Medchemexpress LLC Supplier No. HY113081S11MG
1-Methyladenosine-d3 hydriodide is a deuterium-labeled version of 1-Methyladenosine. This RNA modification serves as a tumor marker, with elevated levels linked to cancer development. Its methylation leads to the upregulation of PPARδ expression, regulating cholesterol metabolism and activating the Hedgehog signaling pathway, contributing to liver tumorigenesis. This compound is intended for research use only and can be used as a tracer and internal standard for quantitative analysis.
  • Deuterium-labeled version of 1-Methyladenosine
  • Functions as a tumor marker
  • Upregulates PPARδ expression
  • Regulates cholesterol metabolism
  • Activates Hedgehog signaling pathway
  • Contributes to liver tumorigenesis
  • Suitable as a tracer for quantitative analysis
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS