Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-methyladenosine-d3 hydrochloride | 00-00-0 | 98.1% | 320.75 | C11H13D3ClN5O4 | 5 MG
SDP

Catalog No. 5000264192
Encompass_Preferred

1-Methyladenosine-d3 hydrochloride is a deuterium-labeled hydrochloride salt used as an isotope-labeled internal standard and research reagent for studying RNA methylation and quantifying methylated nucleosides in biological samples. It provides a stable tracer for mass spectrometry and analytical workflows in epitranscriptomics and biomarker research.

  • Deuterium-labeled internal standard for MS and quantitative assays.
  • Hydrochloride salt form for improved solubility and stability.
  • High purity (98.1%) appropriate for analytical use.
  • Molecular weight 320.75; formula C11H13D3ClN5O4.
  • Available as 5 mg vials for laboratory use.
  • Store at -20°C, sealed, away from moisture and light.

Catalog No. 50-002-64192 Supplier Medchemexpress LLC Supplier No. HY113081AS5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Methyladenosine-d3 hydrochloride is a deuterium-labeled hydrochloride salt used as an isotope-labeled internal standard and research reagent for studying RNA methylation and quantifying methylated nucleosides in biological samples. It provides a stable tracer for mass spectrometry and analytical workflows in epitranscriptomics and biomarker research.

  • Deuterium-labeled internal standard for MS and quantitative assays.
  • Hydrochloride salt form for improved solubility and stability.
  • High purity (98.1%) appropriate for analytical use.
  • Molecular weight 320.75; formula C11H13D3ClN5O4.
  • Available as 5 mg vials for laboratory use.
  • Store at -20°C, sealed, away from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.