Learn More
eMolecules​ 959957-92-7 | 1-METHYLAZETIDIN-3-AMINE | AstaTech | MFCD11849029 | 86.138 | C4H10N2 | 95 | CN1CC(N)C1 | 1g | 268489666
Catalog No. NC2257237 Shop All eMolecules Inc. Products
Encompass_Preferred
1-METHYLAZETIDIN-3-AMINE | AstaTech | 959957-92-7 | MFCD11849029 | 86.138 | C4H10N2 | 95.000 | CN1CC(N)C1 | 1g | 268489666
This item is not returnable. View return policy
Catalog No. NC2257237 Supplier eMolecules​ Supplier No. 268489666
1-METHYLAZETIDIN-3-AMINE | AstaTech | 959957-92-7 | MFCD11849029 | 86.138 | C4H10N2 | 95.000 | CN1CC(N)C1 | 1g | 268489666