Learn More
TARGETMOL CHEMICALS INC 1-METHYLCYTOSINE 10MG
SDP
Catalog No. 502581774
Encompass_Preferred
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1-Methylcytosine (4-amino-1-methylpyrimidin-2(1H)-one) is a methylated form of the cytosine and can be used as the nucleobase of hachimoji DNA paired with isoguanine. purity: 100%
Catalog No. 50-258-1774 Supplier TARGETMOL CHEMICALS INC Supplier No. T1001910MG
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1-Methylcytosine (4-amino-1-methylpyrimidin-2(1H)-one) is a methylated form of the cytosine and can be used as the nucleobase of hachimoji DNA paired with isoguanine. purity: 100%