Learn More
Medchemexpress LLC 1-Methylcytosine | 1122-47-0 | 125.13 | 1 ML
SDP
Catalog No. 5000449316
Encompass_Preferred
1-Methylcytosine is a pyrimidinone that functions as a nucleobase for hachimoji DNA, forming pairs with isoguanine. It exhibits weak antiproliferative activity against cervical cancer cells and is utilized in cervical cancer research.
  • Serves as a nucleobase for hachimoji DNA
  • Pairs with isoguanine
  • Exhibits weak antiproliferative activity against cervical cancer cells
  • Used in the research of cervical cancer
Catalog No. 50-004-49316 Supplier Medchemexpress LLC Supplier No. HYW00639510MM/1ML
1-Methylcytosine is a pyrimidinone that functions as a nucleobase for hachimoji DNA, forming pairs with isoguanine. It exhibits weak antiproliferative activity against cervical cancer cells and is utilized in cervical cancer research.
  • Serves as a nucleobase for hachimoji DNA
  • Pairs with isoguanine
  • Exhibits weak antiproliferative activity against cervical cancer cells
  • Used in the research of cervical cancer