Learn More
Medchemexpress LLC 1-Methylcytosine | 1122-47-0 | 125.13 | 100 MG
SDP
Catalog No. 5000445356
Encompass_Preferred
1-Methylcytosine is a pyrimidinone that acts as a nucleobase for hachimoji DNA, pairing with isoguanine. It demonstrates weak antiproliferative activity against cervical cancer cells and can be utilized in cervical cancer research.
  • Acts as a nucleobase for hachimoji DNA
  • Pairs with isoguanine
  • Exhibits weak antiproliferative activity against cervical cancer cells
  • Suitable for cervical cancer research
Catalog No. 50-004-45356 Supplier Medchemexpress LLC Supplier No. HYW006395100MG
1-Methylcytosine is a pyrimidinone that acts as a nucleobase for hachimoji DNA, pairing with isoguanine. It demonstrates weak antiproliferative activity against cervical cancer cells and can be utilized in cervical cancer research.
  • Acts as a nucleobase for hachimoji DNA
  • Pairs with isoguanine
  • Exhibits weak antiproliferative activity against cervical cancer cells
  • Suitable for cervical cancer research