Learn More
Medchemexpress LLC 1-Methylcytosine | 1122-47-0 | 125.13 | 25 MG
SDP
Catalog No. 5000446683
Encompass_Preferred
1-Methylcytosine is a methylated form of the DNA base cytosine. It is utilized as a nucleobase of hachimoji DNA, where it forms a pair with Isoguanine. It has been observed to exhibit weak antiproliferative activity against cervical cancer cells and can be utilized in cervical cancer research.
  • Weakly inhibits proliferation of human cervical cancer HeLa cells with an IC50 value of 12 μM.
  • Shows no antiproliferative activity against human tumor cell lines such as MiaPaCa-2, SW620, MCF-7, and H460 (IC50 >100 μM for all).
  • Functions as a nucleoside antimetabolite/analog.
  • Involved in cell cycle/DNA damage pathway.
Catalog No. 50-004-46683 Supplier Medchemexpress LLC Supplier No. 25MGHYW006395
1-Methylcytosine is a methylated form of the DNA base cytosine. It is utilized as a nucleobase of hachimoji DNA, where it forms a pair with Isoguanine. It has been observed to exhibit weak antiproliferative activity against cervical cancer cells and can be utilized in cervical cancer research.
  • Weakly inhibits proliferation of human cervical cancer HeLa cells with an IC50 value of 12 μM.
  • Shows no antiproliferative activity against human tumor cell lines such as MiaPaCa-2, SW620, MCF-7, and H460 (IC50 >100 μM for all).
  • Functions as a nucleoside antimetabolite/analog.
  • Involved in cell cycle/DNA damage pathway.