Learn More
Medchemexpress LLC 1-Methylcytosine | 1122-47-0 | 125.13 | 50 MG
SDP
Catalog No. 5000445354
Encompass_Preferred
1-Methylcytosine is a pyrimidinone that acts as a nucleobase for hachimoji DNA, forming a pair with isoguanine. It exhibits weak antiproliferative activity against cervical cancer cells and can be used in cervical cancer research.
  • Acts as a nucleobase for hachimoji DNA
  • Pairs with isoguanine
  • Exhibits weak antiproliferative activity against cervical cancer cells
  • Can be used in cervical cancer research
Catalog No. 50-004-45354 Supplier Medchemexpress LLC Supplier No. HYW00639550MG
1-Methylcytosine is a pyrimidinone that acts as a nucleobase for hachimoji DNA, forming a pair with isoguanine. It exhibits weak antiproliferative activity against cervical cancer cells and can be used in cervical cancer research.
  • Acts as a nucleobase for hachimoji DNA
  • Pairs with isoguanine
  • Exhibits weak antiproliferative activity against cervical cancer cells
  • Can be used in cervical cancer research