Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyldicyclopentadiene 90.0+%, TCI America™
SDP

Catalog No. M0910100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0910100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M0910100MG Supplier TCI America Supplier No. M0910100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16327-42-7
Molecular Formula C11H14
Molecular Weight (g/mol) 146.233
MDL Number MFCD00168054
InChI Key VRLWYYZVFNNMET-UHFFFAOYSA-N
Synonym 5-Methyltricyclo[5.2.1.0(2,6)]deca-3,8-diene, 3a,4,7,7a-Tetrahydro-4,7-methano-1-methyl-1H-indene
PubChem CID 19967616
SMILES CC1C=CC2C1C3CC2C=C3

Specifications

Color Yellow
Quantity 100 mg
UN Number 3295
Formula Weight 146.23
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methyldicyclopentadiene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.