Learn More
Abcam 1-Methylguanine, 250MG
Catalog No. 502671854
Encompass_Preferred
MW 165.15 Da. Naturally occurring modified purine derived from tRNA, found in elevated levels in the serum and urine of cancer patients. (PMID: 2413515).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1854 Supplier Abcam Supplier No. AB275320250MG
MW 165.15 Da. Naturally occurring modified purine derived from tRNA, found in elevated levels in the serum and urine of cancer patients. (PMID: 2413515).
The product is subject to the following: Abcam Restricted Use Statement