Learn More
Medchemexpress LLC 1-methylhistamine-d3 dihydrochloride | 344299-49-6 | 99.9% | 201.11 g/mol | C6H10D3Cl2N3 | 1 MG
SDP
Catalog No. 5000209951
Encompass_Preferred
1-Methylhistamine-d3 dihydrochloride is the deuterium-labeled form of 1-methylhistamine supplied as the dihydrochloride salt. It is used as a stable isotope internal standard or tracer in analytical workflows such as LC-MS and metabolomics.
  • Deuterium-labeled (d3) internal standard for quantitative analysis.
  • Supplied as the dihydrochloride salt for stability and handling.
  • High purity suitable for analytical and metabolomics applications.
  • Common applications include tracer studies and internal standardization in LC-MS assays.
  • Available in small quantities appropriate for standards and calibration.
Catalog No. 50-002-09951 Supplier Medchemexpress LLC Supplier No. HYW053787S1MG
1-Methylhistamine-d3 dihydrochloride is the deuterium-labeled form of 1-methylhistamine supplied as the dihydrochloride salt. It is used as a stable isotope internal standard or tracer in analytical workflows such as LC-MS and metabolomics.
  • Deuterium-labeled (d3) internal standard for quantitative analysis.
  • Supplied as the dihydrochloride salt for stability and handling.
  • High purity suitable for analytical and metabolomics applications.
  • Common applications include tracer studies and internal standardization in LC-MS assays.
  • Available in small quantities appropriate for standards and calibration.