Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-methylhistamine dihydrochloride | 6481-48-7 | 99.9% | 198.09 g/mol | C6H13Cl2N3 | 1 ML
SDP

Catalog No. 5000210041
Encompass_Preferred

Ready-to-use 10 mM solution of 1-methylhistamine dihydrochloride in water (1 mL) supplied as an analytical standard for research use. Common applications include quantitative analysis, method validation, and use as an endogenous metabolite reference. Store sealed at 4°C, protected from moisture.

  • Ready-to-use 10 mM aqueous standard, 1 mL
  • High purity 99.9% as reported on the COA
  • Molecular weight 198.09 g/mol and formula C6H13Cl2N3
  • Suitable for quantitative analysis and method validation
  • Store at 4°C, sealed and away from moisture

Catalog No. 50-002-10041 Supplier Medchemexpress LLC Supplier No. HYW05378710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready-to-use 10 mM solution of 1-methylhistamine dihydrochloride in water (1 mL) supplied as an analytical standard for research use. Common applications include quantitative analysis, method validation, and use as an endogenous metabolite reference. Store sealed at 4°C, protected from moisture.

  • Ready-to-use 10 mM aqueous standard, 1 mL
  • High purity 99.9% as reported on the COA
  • Molecular weight 198.09 g/mol and formula C6H13Cl2N3
  • Suitable for quantitative analysis and method validation
  • Store at 4°C, sealed and away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.