Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-methylhistamine dihydrochloride | 6481-48-7 | MFCD00037000 | 99.9% | 198.09 g/mol | C6H13Cl2N3 | 5 MG
SDP

Catalog No. 5000209939
Encompass_Preferred

1-Methylhistamine dihydrochloride is an endogenous histamine metabolite supplied as an analytical standard for research and reference use. It is provided as a solid powder with high purity for use in biochemical and analytical assays.

  • Purity: 99.9%.
  • Physical form: solid, off-white to gray.
  • Molecular weight: 198.09 g/mol; chemical formula: C6H13Cl2N3.
  • CAS number: 6481-48-7.
  • Pack size: 5 mg (other sizes available).
  • Storage: store sealed at 4°C, away from moisture; in solution keep at -80°C up to 6 months or -20°C up to 1 month.
  • Use: for research use only; not for human or clinical use.

Catalog No. 50-002-09939 Supplier Medchemexpress LLC Supplier No. HYW0537875MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Methylhistamine dihydrochloride is an endogenous histamine metabolite supplied as an analytical standard for research and reference use. It is provided as a solid powder with high purity for use in biochemical and analytical assays.

  • Purity: 99.9%.
  • Physical form: solid, off-white to gray.
  • Molecular weight: 198.09 g/mol; chemical formula: C6H13Cl2N3.
  • CAS number: 6481-48-7.
  • Pack size: 5 mg (other sizes available).
  • Storage: store sealed at 4°C, away from moisture; in solution keep at -80°C up to 6 months or -20°C up to 1 month.
  • Use: for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.