Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Methylhomopiperazine

Catalog No. 1860905ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-609-05ML 5 mL
18-609-025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-609-05ML Supplier Sigma Aldrich Supplier No. 1860905ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-methylhomopiperazine undergoes coupling reaction with a series of diazonium salts to afford the 4-methyl-1-[2-aryl-1-diazenyl]-1,4-diazepanes.1-methylhomopiperazine was used in the synthesis of:
  • Novel benzimidazole derivatives having pharmacological activities
  • Platinum(ii) substituted disulfide complexes

Specifications

CAS 4318-37-0
Density 0.918 g/mL (at 25°C (literature))
Boiling Point 74°C to 75°C (35 mmHg)
Molecular Formula C6H14N2
Refractive Index n20/D 1.477 (literature)
Linear Formula C6H14N2
MDL Number MFCD00059810
Quantity 5 mL
Synonym Hexahydro-1-methyl-1 H-1,4-diazepine
Molecular Weight (g/mol) 114.19
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.