Learn More
Medchemexpress LLC 1-Methylimidazole | 616-47-7 | 99.98% | C4H6N2 | 1 KG
SDP
Catalog No. 5000433987
Encompass_Preferred
This product is an imidazole derivative primarily utilized as a ligand for ruthenium (II) complexes, specifically in research pertaining to non-small cell lung cancer studies. It presents as a colorless to light yellow liquid with a density of 1.035 g/cm³ and boasts a high purity of 99.98%. For laboratory applications, it exhibits excellent solubility in DMSO at 200 mg/mL, with ultrasonic assistance recommended for optimal dissolution.
  • Three years shelf life at room temperature
  • Two years shelf life when stored in solvent at -80°C
  • One year shelf life when stored in solvent at -20°C
Catalog No. 50-004-33987 Supplier Medchemexpress LLC Supplier No. HY591371KG
This product is an imidazole derivative primarily utilized as a ligand for ruthenium (II) complexes, specifically in research pertaining to non-small cell lung cancer studies. It presents as a colorless to light yellow liquid with a density of 1.035 g/cm³ and boasts a high purity of 99.98%. For laboratory applications, it exhibits excellent solubility in DMSO at 200 mg/mL, with ultrasonic assistance recommended for optimal dissolution.
  • Three years shelf life at room temperature
  • Two years shelf life when stored in solvent at -80°C
  • One year shelf life when stored in solvent at -20°C