Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Methylimidazole | 616-47-7 | 99.98% | C4H6N2 | 500 G
SDP

Catalog No. 5000434197
Encompass_Preferred

1-Methylimidazole can be used as a ligand for ruthenium (II) complexes in the study of non-small cell lung cancer. It is for research use only.

  • Used as a ligand for ruthenium (II) complexes
  • For research use only
  • Molecular weight: 82.10
  • Appearance: Colorless to light yellow liquid (Density: 1.035 g/cm³)
  • Related classifications include Cancer, Cancer Metabolism and Metastasis, and Drug Intermediate

Catalog No. 50-004-34197 Supplier Medchemexpress LLC Supplier No. HY59137500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Methylimidazole can be used as a ligand for ruthenium (II) complexes in the study of non-small cell lung cancer. It is for research use only.

  • Used as a ligand for ruthenium (II) complexes
  • For research use only
  • Molecular weight: 82.10
  • Appearance: Colorless to light yellow liquid (Density: 1.035 g/cm³)
  • Related classifications include Cancer, Cancer Metabolism and Metastasis, and Drug Intermediate

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.