Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Methylimidazole | 616-47-7 | MFCD00005292 | 99.99% | C4H6N2 | 100 G
SDP

Catalog No. 5000433785
Encompass_Preferred

1-Methylimidazole is a chemical compound that acts as a ligand for ruthenium (II) complexes, primarily investigated in the context of non-small cell lung cancer research. This product is strictly for research use only and is not intended for patient use.

  • Functions as a ligand for ruthenium (II) complexes
  • Explored in the study of non-small cell lung cancer
  • Provided as a colorless to light yellow liquid
  • Molecular formula is C4H6N2
  • Molecular weight is 82.10
  • High purity of 99.99%
  • Chemical has a CAS number of 616-47-7

Catalog No. 50-004-33785 Supplier Medchemexpress LLC Supplier No. HY59137100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Methylimidazole is a chemical compound that acts as a ligand for ruthenium (II) complexes, primarily investigated in the context of non-small cell lung cancer research. This product is strictly for research use only and is not intended for patient use.

  • Functions as a ligand for ruthenium (II) complexes
  • Explored in the study of non-small cell lung cancer
  • Provided as a colorless to light yellow liquid
  • Molecular formula is C4H6N2
  • Molecular weight is 82.10
  • High purity of 99.99%
  • Chemical has a CAS number of 616-47-7

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.