Learn More
Medchemexpress LLC 1-methylimidazoleacetic acid (standard) | 2625-49-2 | 140.14 | 250 MG
SDP
Catalog No. 5000348873
Encompass_Preferred
1-Methylimidazoleacetic acid (Standard) is the analytical standard for 1-Methylimidazoleacetic acid. This compound, also known as 1-Methylimidazole-4-acetic acid, is an endogenous metabolite of histamine found in urine, making it useful in the study of mastocytosis. It is intended for research and analytical applications, particularly in qualitative, quantitative, and methodological research experiments.
  • Analytical standard
  • Intended for research and analytical applications
  • Histamine metabolite in urine, useful in mastocytosis studies
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
  • Reference standard supplied assay
Catalog No. 50-003-48873 Supplier Medchemexpress LLC Supplier No. 250MGHY13936
1-Methylimidazoleacetic acid (Standard) is the analytical standard for 1-Methylimidazoleacetic acid. This compound, also known as 1-Methylimidazole-4-acetic acid, is an endogenous metabolite of histamine found in urine, making it useful in the study of mastocytosis. It is intended for research and analytical applications, particularly in qualitative, quantitative, and methodological research experiments.
  • Analytical standard
  • Intended for research and analytical applications
  • Histamine metabolite in urine, useful in mastocytosis studies
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
  • Reference standard supplied assay