Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Methylimidazolium Hydrogen Sulfate

Catalog No. 597601KGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-760-1KGF 1 kg
59-760-100GF 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 59-760-1KGF Supplier Sigma Aldrich Supplier No. 597601KGF
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Methylimidazolium hydrogen sulfate is a Br++nsted acidic ionic liquid that can be used:As a component of a catalytic system along with chlorotrimethylsilane used in the preparation of 3,4-dihydropyrimidin-2(1H)-ones and hydroquinazoline-2,5-diones.As a catalyst in the preparation of biologically important 2,5-dimethyl-N-substituted pyrroles.As a reusable catalyst in the preparation of spiro[indoline-3'sa,2-quinazoline]2'sa,4(3H)-dione.In the leaching of copper and zinc in brass dross using H2O2 as an oxidant.

Specifications

CAS 681281-87-8
Molecular Formula C4H6N2 -+ H2SO4
Linear Formula C4H6N2 -+ H2SO4
MDL Number MFCD06798198
Quantity 1 kg
Synonym 1-Methylimidazole hydrogen sulfate
Molecular Weight (g/mol) 180.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.