Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylindazole-3-carboxylic acid, 97%

Catalog No. AAH2687903
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAH2687903

Supplier: Thermo Scientific Chemicals H2687903

Only null left
Add to Cart

CAS: 50890-83-0 | C9H8N2O2 | 176.175 g/mol

1-Methylindazole-3-carboxylic acid is used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a pharmaceutical adjuvant, Granisetron Impurity D.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylindazole-3-carboxylic acid is used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a pharmaceutical adjuvant, Granisetron Impurity D.

Solubility
Insoluble in water. Soluble in DMSO and Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50890-83-0
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD00272569
InChI Key OVVDFORZEGKEJM-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazole-3-carboxylic acid, n-methyl-indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylic acid, 1h-indazole-3-carboxylic acid, 1-methyl, 1-methyl-indazole-3-carboxylic acid, n-methylindazole-3-carboxylic acid, 1-methylindozole-3-carboxylic acid, n-methyl-indozole-3-carboxylic acid, n-methyl-1h indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylicacid
PubChem CID 689105
IUPAC Name 1-methylindazole-3-carboxylic acid
SMILES CN1C2=CC=CC=C2C(=N1)C(=O)O

Specifications

Melting Point ∽218°C (decomposition)
Quantity 1 g
Solubility Information Insoluble in water. Soluble in DMSO and Methanol.
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material 1-Methylindazole-3-carboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.