Learn More
Medchemexpress LLC 1-Methylinosine | 2140-73-0 | 99.7% | 282.25 | 50 MG
SDP
Catalog No. 501969230
Encompass_Preferred
1-Methylinosine is a modified nucleotide that plays a role in eukaryotic tRNA, specifically at position 37. As a metabolite derived from inosine, its elevated levels have been observed in urine samples from breast cancer models.
  • Modified nucleotide for eukaryotic tRNA
  • Metabolite derived from inosine
  • Elevated levels observed in breast cancer models
  • White to off-white solid appearance
Catalog No. 50-196-9230 Supplier Medchemexpress LLC Supplier No. HY11313950MG
1-Methylinosine is a modified nucleotide that plays a role in eukaryotic tRNA, specifically at position 37. As a metabolite derived from inosine, its elevated levels have been observed in urine samples from breast cancer models.
  • Modified nucleotide for eukaryotic tRNA
  • Metabolite derived from inosine
  • Elevated levels observed in breast cancer models
  • White to off-white solid appearance