Learn More
Medchemexpress LLC 1-methylinosine-d3 | 00-00-0 | >98.0% | 285.27 g/mol | C11H11D3N4O5 | 5 MG
SDP
Catalog No. 5000262975
Encompass_Preferred
1-Methylinosine-d3 is the deuterium-labeled isotopologue of 1-methylinosine, a modified nucleoside located at position 37 (3' to the anticodon) of eukaryotic tRNA. Supplied as a stable isotope standard for analytical and research applications, it is provided in small milligram quantities and characterized for high purity.
Catalog No. 50-002-62975 Supplier Medchemexpress LLC Supplier No. HY113139S5MG
1-Methylinosine-d3 is the deuterium-labeled isotopologue of 1-methylinosine, a modified nucleoside located at position 37 (3' to the anticodon) of eukaryotic tRNA. Supplied as a stable isotope standard for analytical and research applications, it is provided in small milligram quantities and characterized for high purity.